Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153310.2 | NP_695222.2 | 5, 6 | 416 | ||
XM_008763489.1 | XP_008761711.1 | 5, 6 | 971 | |||
XM_008763490.1 | XP_008761712.1 | 6, 7 | 1632 | |||
XM_008763491.1 | XP_008761713.1 | 5, 6 | 1402 | |||
XM_008763492.1 | XP_008761714.1 | 5, 6 | 1476 | |||
XM_008763493.1 | XP_008761715.1 | 4, 5 | 334 | |||
GenBank mRNA | AF031942.1 | - | 2, 3 | 240 | ||
U67080.1 | - | 5, 6 | 429 |