Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153310.2 | NP_695222.2 | 6 | 832 | ||
XM_008763489.1 | XP_008761711.1 | 6 | 1387 | |||
XM_008763490.1 | XP_008761712.1 | 7 | 2048 | |||
XM_008763491.1 | XP_008761713.1 | 6 | 1818 | |||
XM_008763492.1 | XP_008761714.1 | 6 | 1892 | |||
XM_008763493.1 | XP_008761715.1 | 5 | 750 | |||
XM_008763494.1 | XP_008761716.1 | 2 | 534 | |||
GenBank mRNA | AF031942.1 | - | 3 | 656 | ||
U67080.1 | - | 6 | 845 |