Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153311.1 | NP_695223.1 | 7 | 675 | ||
XM_008766194.1 | XP_008764416.1 | 5 | 538 | |||
GenBank mRNA | AF537098.1 | - | 7 | 675 | ||
AF537099.1 | - | 5 | 453 |