Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153476.1 | NP_703206.1 | 5, 6 | 1153 | ||
GenBank mRNA | AY128946.1 | - | 5, 6 | 1153 |