Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173116.1 | NP_775139.1 | 2, 3 | 150 | ||
XM_008772870.1 | XP_008771092.1 | 2, 3 | 89 | |||
XM_008772871.1 | XP_008771093.1 | 2, 3 | 99 | |||
XM_008772872.1 | XP_008771094.1 | 1, 2 | 70 | |||
XM_008772873.1 | XP_008771095.1 | 2, 3 | 157 | |||
GenBank mRNA | AJ512838.1 | - | 2, 3 | 150 |