Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246843.2 | XP_006246905.1 | 4 | 1748 | ||
XM_006246844.2 | XP_006246906.1 | 3 | 1572 |