Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175579.3 | NP_783169.2 | 2 | 768 | ||
XM_006247278.2 | XP_006247340.1 | 2 | 873 | |||
XM_006247279.2 | XP_006247341.1 | 2 | 873 | |||
XM_006247280.2 | XP_006247342.1 | 3 | 280 | |||
GenBank mRNA | AY187039.1 | - | 2 | 745 | ||
AY192567.1 | - | 2 | 768 |