Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768541.1 | XP_008766763.1 | 8 | 2031 | ||
XM_237855.8 | XP_237855.6 | 8 | 2031 |