Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105909.1 | NP_001099379.1 | 8 | 905 | ||
XM_008768948.1 | XP_008767170.1 | 8 | 915 |