Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769281.1 | XP_008767503.1 | 20 | 5832 | ||
XM_008769282.1 | XP_008767504.1 | 20 | 5826 | |||
XM_008769285.1 | XP_008767507.1 | 20 | 5831 | |||
XM_008769286.1 | XP_008767508.1 | 20 | 5825 | |||
XM_008769287.1 | XP_008767509.1 | 20 | 5832 | |||
XM_008769289.1 | XP_008767511.1 | 20 | 5832 | |||
XM_008769290.1 | XP_008767512.1 | 20 | 5831 | |||
XM_008769292.1 | XP_008767514.1 | 19 | 5622 | |||
XM_008769294.1 | XP_008767516.1 | 20 | 5832 | |||
XM_008769295.1 | XP_008767517.1 | 20 | 6056 | |||
GenBank mRNA | FQ230562.1 | - | 3 | 299 |