Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105980.1 | NP_001099450.1 | 8 | 843 | ||
XM_006250367.2 | XP_006250429.1 | 8 | 767 | |||
XM_006250368.2 | XP_006250430.1 | 8 | 897 | |||
XM_006250370.2 | XP_006250432.1 | 4 | 476 | |||
XM_006250372.2 | XP_006250434.1 | 4 | 350 |