Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106103.1 | NP_001099573.1 | 7 | 920 | ||
XM_006253696.2 | XP_006253758.2 | 8 | 1366 | |||
XM_006253699.2 | XP_006253761.1 | 8 | 1366 | |||
XM_008771490.1 | XP_008769712.1 | 8 | 1366 | |||
XM_008771558.1 | XP_008769780.1 | 8 | 1366 | |||
GenBank mRNA | FQ221052.1 | - | 6 | 832 |