Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008766.1 | NP_001008766.1 | 1, 2 | 223 | ||
GenBank mRNA | BC088474.1 | - | 1, 2 | 223 | 7129030 | |
FQ214368.1 | - | 1, 2 | 246 | |||
FQ230934.1 | - | 1, 2 | 237 | |||
FQ231672.1 | - | 1, 2 | 216 |