Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107534.1 | NP_001101004.1 | 18 | 2928 | ||
GenBank mRNA | FQ221869.1 | - | 11 | 2136 |