Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039849.2 | NP_001034938.1 | 3 | 459 | ||
NM_017165.2 | NP_058861.3 | 3 | 345 | |||
GenBank mRNA | AF274028.1 | - | 3 | 468 | ||
AY570513.1 | - | 3 | 447 | |||
CB582508.1 | - | 3 | 369 | |||
FQ210610.1 | - | 3 | 253 | |||
FQ219811.1 | - | 3 | 253 | |||
FQ221663.1 | - | 3 | 275 | |||
FQ221760.1 | - | 3 | 253 | |||
FQ221899.1 | - | 3 | 253 | |||
FQ221976.1 | - | 3 | 256 | |||
FQ222299.1 | - | 3 | 253 | |||
FQ227564.1 | - | 3 | 241 | |||
FQ227764.1 | - | 3 | 412 | |||
FQ231108.1 | - | 3 | 311 | |||
FQ231686.1 | - | 3 | 253 | |||
FQ233099.1 | - | 3 | 253 | |||
FQ234630.1 | - | 3 | 253 | |||
L24896.1 | - | 3 | 222 | |||
U37427.1 | - | 3 | 339 | |||
X82679.1 | - | 3 | 232 |