Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113357.1 | NP_001106828.1 | 2 | 340 | ||
XM_006253024.2 | XP_006253086.1 | 3 | 735 | |||
XM_006253025.2 | XP_006253087.1 | 3 | 648 | |||
GenBank mRNA | Z11504.1 | - | 1 | 235 |
Set Membership: |
![]() |