Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277272.1 | NP_001264201.1 | 1118 | |||
XM_006229555.2 | XP_006229617.1 | 1607 | ||||
GenBank mRNA | BC103640.1 | - | 1476 | 7134953 | ||
BC158688.1 | - | 1050 | 8375819 |