Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106381.1 | NP_001099851.1 | 10 | 1663 | ||
XM_006256338.2 | XP_006256400.1 | 10 | 1683 | |||
GenBank mRNA | BC168843.1 | - | 9 | 1455 | 9089070 |