Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177321.1 | NP_001170792.1 | 7 | 719 | ||
NM_017200.1 | NP_058896.1 | 6 | 647 | |||
XM_006234440.2 | XP_006234502.1 | 7 | 942 | |||
XM_006234441.2 | XP_006234503.1 | 7 | 1963 | |||
XM_006234444.2 | XP_006234506.1 | 7 | 1976 | |||
XM_008761980.1 | XP_008760202.1 | 7 | 961 | |||
XM_008761981.1 | XP_008760203.1 | 7 | 725 | |||
XM_008761982.1 | XP_008760204.1 | 7 | 955 | |||
XM_008761983.1 | XP_008760205.1 | 7 | 1978 | |||
XM_008761984.1 | XP_008760206.1 | 7 | 1991 | |||
XM_008761985.1 | XP_008760207.1 | 7 | 967 | |||
XM_008761986.1 | XP_008760208.1 | 7 | 961 | |||
GenBank mRNA | BC087020.1 | - | 7 | 719 | 7192382 | |
D10926.1 | - | 6 | 647 | |||
FQ219876.1 | - | 4 | 353 | |||
FQ220339.1 | - | 4 | 347 | |||
FQ220655.1 | - | 4 | 444 | |||
FQ220711.1 | - | 4 | 444 | |||
FQ229364.1 | - | 4 | 376 |