Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106408.2 | NP_001099878.2 | 7, 8 | 724 | ||
XM_006231902.2 | XP_006231964.1 | 7, 8 | 752 | |||
XM_006231903.2 | XP_006231965.1 | 8, 9 | 935 | |||
XM_006231905.2 | XP_006231967.1 | 6, 7 | 638 | |||
GenBank mRNA | BC158729.1 | - | 7, 8 | 724 | 9091721 |