Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106465.1 | NP_001099935.1 | 3 | 1214 | ||
XM_008761393.1 | XP_008759615.1 | 1 | 380 | |||
XM_008761394.1 | XP_008759616.1 | 1 | 380 |