Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Htr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017254.1 | NP_058950.1 | 3 | 1375 | ||
GenBank mRNA | M30705.1 | - | 3 | 1375 | ||
M64867.1 | - | 1781 | ||||
X13971.1 | - | 1918 |
Set Membership: |
G-protein coupled receptor |