Gene Symbol: Emc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106495.1 | NP_001099965.1 | 2 | 249 | ||
XM_006234704.2 | XP_006234766.1 | 2 | 1514 | |||
XM_008762088.1 | XP_008760310.1 | 2 | 1509 | |||
GenBank mRNA | FQ215850.1 | - | 2 | 189 | ||
FQ234005.1 | - | 2 | 189 |