Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106574.1 | NP_001100044.1 | 2 | 1654 | ||
XM_006235890.2 | XP_006235952.1 | 3 | 811 | |||
XM_006235891.2 | XP_006235953.1 | 2 | 159 | |||
XM_008762633.1 | XP_008760855.1 | 3 | 833 |