Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mcm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017287.1 | NP_058983.1 | 16 | 2399 | ||
GenBank mRNA | BC166822.1 | - | 16 | 2319 | 8369193 | |
BQ194934.1 | - | 2 | 516 | |||
CF109502.1 | - | 5 | 665 | |||
U17565.1 | - | 10 | 1349 |