Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022710.1 | NP_073201.1 | 3 | 200 | ||
XM_006242386.2 | XP_006242448.1 | 2 | 772 | |||
GenBank mRNA | AF327905.1 | - | 3 | 156 | ||
AF327906.1 | - | 3 | 193 | |||
FQ212460.1 | - | 3 | 256 | |||
M94537.1 | - | 3 | 200 |