Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270772.1 | NP_001257701.1 | 1 | 387 | ||
NM_001270773.1 | NP_001257702.1 | 1 | 387 | |||
NM_001270774.1 | NP_001257703.1 | 1 | 387 | |||
NM_001270775.1 | NP_001257704.1 | 1 | 387 | |||
NM_001270776.1 | NP_001257705.1 | 1 | 387 | |||
NM_001270777.1 | NP_001257706.1 | 1 | 387 | |||
NM_001270778.1 | NP_001257707.1 | 1 | 387 | |||
NM_001270779.1 | NP_001257708.1 | 1 | 387 | |||
NM_019268.3 | NP_062141.3 | 1 | 387 | |||
XM_006239660.2 | XP_006239722.1 | 2 | 904 | |||
XM_008764432.1 | XP_008762654.1 | 2 | 551 | |||
XM_008764433.1 | XP_008762655.1 | 2 | 551 | |||
XM_008764434.1 | XP_008762656.1 | 2 | 550 | |||
XM_008764435.1 | XP_008762657.1 | 2 | 551 | |||
XM_008764436.1 | XP_008762658.1 | 2 | 550 | |||
XM_008764437.1 | XP_008762659.1 | 2 | 550 | |||
XM_008764438.1 | XP_008762660.1 | 2 | 550 | |||
XM_008764439.1 | XP_008762661.1 | 2 | 550 | |||
GenBank mRNA | AF109163.1 | - | 1 | 353 | ||
AF109164.1 | - | 1 | 353 | |||
AF109165.1 | - | 1 | 353 | |||
AF109166.1 | - | 1 | 353 | |||
AY033398.1 | - | 1 | 353 | |||
BC078698.1 | - | 711 | ||||
U04933.1 | - | 1 | 729 | |||
X68191.1 | - | 1 | 416 | |||
X68812.1 | - | 1 | 467 | |||
X68813.1 | - | 1 | 478 |