Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017308.1 | NP_059004.1 | 6, 7 | 576 | ||
XM_006232858.2 | XP_006232920.1 | 9, 10 | 988 | |||
XM_006232861.2 | XP_006232923.1 | 7, 8 | 850 | |||
XM_006232869.2 | XP_006232931.1 | 5, 6 | 497 | |||
XM_008761286.1 | XP_008759508.1 | 7, 8 | 4254 | |||
XM_008761287.1 | XP_008759509.1 | 7, 8 | 3601 | |||
XM_008761288.1 | XP_008759510.1 | 7, 8 | 2527 | |||
XM_008761289.1 | XP_008759511.1 | 7, 8 | 4106 | |||
XM_008761290.1 | XP_008759512.1 | 7, 8 | 3353 | |||
XM_008761291.1 | XP_008759513.1 | 7, 8 | 3684 | |||
XM_008761292.1 | XP_008759514.1 | 7, 8 | 2783 | |||
XM_008761293.1 | XP_008759515.1 | 8, 9 | 940 | |||
XM_008761294.1 | XP_008759516.1 | 6, 7 | 4412 | |||
GenBank mRNA | AB000817.2 | - | 6, 7 | 592 | ||
AB014074.1 | - | 6, 7 | 592 |