Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106692.1 | NP_001100162.1 | 3 | 190 | ||
XM_008764220.1 | XP_008762442.1 | 3 | 198 | |||
GenBank mRNA | FQ231865.1 | - | 3 | 170 |