Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106713.2 | NP_001100183.1 | 11 | 2755 | ||
XM_006239857.2 | XP_006239919.1 | 11 | 4363 | |||
XM_008764550.1 | XP_008762772.1 | 10 | 5951 |