Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765471.1 | XP_008763693.1 | 1407 | |||
XM_008765472.1 | XP_008763694.1 | 1407 | ||||
XM_008765473.1 | XP_008763695.1 | 1407 |