Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765471.1 | XP_008763693.1 | 1117 | |||
XM_008765472.1 | XP_008763694.1 | 1117 | ||||
XM_008765473.1 | XP_008763695.1 | 1117 | ||||
XM_008765474.1 | XP_008763696.1 | 1117 |
Gene Symbol: Dcstamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235262.8 | XP_235262.4 | 1 | 1117 |