Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134469.1 | NP_001127941.1 | 7 | 675 | ||
XM_006243202.2 | XP_006243264.1 | 8 | 1147 | |||
XM_006243204.2 | XP_006243266.1 | 7 | 1091 | |||
GenBank mRNA | BC160848.1 | - | 7 | 675 | 8363379 | |
FQ211943.1 | - | 6 | 828 |