Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106853.1 | NP_001100323.1 | 9 | 1009 | ||
XM_006243728.2 | XP_006243790.1 | 10 | 1299 | |||
XM_006243729.2 | XP_006243791.1 | 10 | 1619 | |||
XM_006243730.2 | XP_006243792.1 | 9 | 1374 | |||
XM_008766502.1 | XP_008764724.1 | 10 | 1234 | |||
GenBank mRNA | BC161944.1 | - | 9 | 1003 | 9024451 |