Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LTF SIRNA Assays ›
Gene Symbol:
Ltf
Gene Name:
lactotransferrin
Gene Aliases:
-
Chromosome Location:
Chr.8: 119290416 - 119313261 on Build Rnor_6.0
UniGene ID:
Rn.218647
Species:
Rat
siRNA ID
RSS313106
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ltf
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001106864.1
NP_001100334.1
10
1273
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytokine production
retina homeostasis
innate immune response in mucosa
proteolysis
iron ion transport
cellular iron ion homeostasis
biological_process
antibacterial humoral response
antifungal humoral response
negative regulation of lipopolysaccharide-mediated signaling pathway
regulation of tumor necrosis factor production
negative regulation of ATPase activity
positive regulation of osteoblast proliferation
positive regulation of toll-like receptor 4 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of viral genome replication
positive regulation of osteoblast differentiation
negative regulation of viral process
positive regulation of NF-kappaB transcription factor activity
bone morphogenesis
positive regulation of protein serine/threonine kinase activity
negative regulation of single-species biofilm formation in or on host organism
positive regulation of chondrocyte proliferation
molecular_function
iron ion binding
ferric iron binding
heparin binding
peptidase activity
protein serine/threonine kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0