Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106883.1 | NP_001100353.1 | 2 | 423 | ||
GenBank mRNA | BC168177.1 | - | 2 | 410 | 9035251 | |
FQ214885.1 | - | 2 | 425 | |||
FQ214919.1 | - | 2 | 315 | |||
FQ215260.1 | - | 2 | 425 | |||
FQ215792.1 | - | 2 | 425 | |||
FQ216053.1 | - | 2 | 425 | |||
FQ216174.1 | - | 2 | 314 | |||
FQ216724.1 | - | 2 | 422 | |||
FQ216808.1 | - | 2 | 311 | |||
FQ217810.1 | - | 2 | 425 | |||
FQ224967.1 | - | 2 | 458 |