Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039020.1 | NP_001034109.1 | 11 | 1382 | ||
XM_006247001.1 | XP_006247063.1 | 12 | 1536 | |||
XM_006247002.1 | XP_006247064.1 | 12 | 1536 | |||
XM_008768060.1 | XP_008766282.1 | 12 | 1536 | |||
XM_008768061.1 | XP_008766283.1 | 12 | 1536 | |||
XM_008768062.1 | XP_008766284.1 | 11 | 1488 | |||
GenBank mRNA | BC100269.1 | - | 11 | 1382 | 7387460 | |
FQ221862.1 | - | 9 | 1703 |