Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107142.1 | NP_001100612.1 | 9 | 1294 | ||
XM_006249365.1 | XP_006249427.1 | 8 | 1212 |