- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Pole | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107152.2 | NP_001100622.2 | 25, 26 | 3113 | ||
XM_006249562.2 | XP_006249624.1 | 25, 26 | 3150 | |||
XM_008769335.1 | XP_008767557.1 | 25, 26 | 3150 |