Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107165.1 | NP_001100635.1 | 5 | 1019 | ||
XM_008772381.1 | XP_008770603.1 | 6 | 1143 | |||
XM_008772382.1 | XP_008770604.1 | 6 | 1158 | |||
XM_008772383.1 | XP_008770605.1 | 6 | 1140 | |||
XM_008772384.1 | XP_008770606.1 | 6 | 1117 | |||
XM_008772385.1 | XP_008770607.1 | 7 | 1079 | |||
XM_008772386.1 | XP_008770608.1 | 7 | 954 | |||
XM_008772387.1 | XP_008770609.1 | 6 | 972 | |||
XM_008772388.1 | XP_008770610.1 | 6 | 1089 | |||
XM_008772389.1 | XP_008770611.1 | 6 | 1066 | |||
XM_008772390.1 | XP_008770612.1 | 6 | 1085 | |||
XM_008772391.1 | XP_008770613.1 | 6 | 1070 | |||
XM_008772392.1 | XP_008770614.1 | 6 | 784 | |||
XM_008772393.1 | XP_008770615.1 | 6 | 1093 | |||
XM_008772394.1 | XP_008770616.1 | 6 | 873 | |||
XM_008772395.1 | XP_008770617.1 | 6 | 866 | |||
XM_008772396.1 | XP_008770618.1 | 6 | 946 | |||
XM_008772397.1 | XP_008770619.1 | 6 | 899 | |||
XM_008772398.1 | XP_008770620.1 | 6 | 847 |