Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199463.2 | NP_955750.1 | 7, 8 | 2020 | ||
XM_006249774.2 | XP_006249836.1 | 7, 8 | 1870 | |||
GenBank mRNA | AY208851.1 | - | 7, 8 | 1951 | ||
AY429675.1 | - | 7, 8 | 2020 |