Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012026.1 | NP_001012026.1 | 11 | 1456 | ||
XM_006249777.2 | XP_006249839.1 | 11 | 1517 | |||
XM_006249778.2 | XP_006249840.1 | 10 | 1374 | |||
XM_006249779.2 | XP_006249841.1 | 10 | 1371 | |||
GenBank mRNA | BC083647.1 | - | 11 | 1456 | 7135053 |