Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107273.1 | NP_001100743.1 | 15 | 2318 | ||
XM_006252117.2 | XP_006252179.1 | 15 | 2395 | |||
XM_006252118.2 | XP_006252180.1 | 15 | 2503 | |||
XM_006252120.2 | XP_006252182.1 | 15 | 2392 | |||
XM_006252121.2 | XP_006252183.1 | 12 | 2449 | |||
XM_008770757.1 | XP_008768979.1 | 12 | 1775 | |||
GenBank mRNA | FQ211026.1 | - | 15 | 2302 |