Gene Symbol: RGD1308601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107374.1 | NP_001100844.1 | 1 | 341 | ||
XM_006254964.2 | XP_006255026.1 | 1 | 485 |
Gene Symbol: LOC100909715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752206.3 | XP_003752254.1 | 1 | 481 |