Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107406.1 | NP_001100876.1 | 3 | 391 | ||
XM_008772151.1 | XP_008770373.1 | 2 | 604 |