Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025000.1 | NP_001020171.1 | 3, 4 | 534 | ||
GenBank mRNA | BC091241.1 | - | 3, 4 | 534 | 7326298 |