Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103689976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758866.1 | XP_008757088.1 | 1 | 1270 |
Gene Symbol: LOC103690981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758798.1 | XP_008757020.1 | 2 | 1822 |
Gene Symbol: RGD1560718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758795.1 | XP_008757017.1 | 2 | 1473 |
Gene Symbol: Smok2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191619.1 | NP_001178548.1 | 1267 | |||
XM_574319.4 | XP_574319.2 | 1 | 1368 |