Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201562.3 | NP_963856.1 | 3 | 275 | ||
GenBank mRNA | AY279211.1 | - | 3 | 275 | ||
BC129079.1 | - | 3 | 390 | 8370402 |