Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730283.4 | XP_002730329.2 | 7 | 728 | ||
XM_006257244.2 | XP_006257306.1 | 7 | 648 | |||
XM_006257245.2 | XP_006257307.1 | 7 | 641 |