Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009688.1 | NP_001009688.1 | 4 | 857 | ||
GenBank mRNA | BC087659.1 | - | 4 | 857 | 7314010 |